Learn More
Hygiena SnapShot™ ATP Devices
Sensitive, accurate, and reliable single-shot device
Manufacturer:  HYGIENA LLC SBC1575
 View more versions of this product
Catalog No. 22-0377-53
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
2° to 8°C
BioControl Lightning and MVP luminometers
Safety and Handling
ShelfLife : 15 Months