Hygiena SnapShot™ Universal ATP Surface Test

Designed to be compatible with luminometers of most manufacturers

Manufacturer: HYGIENA LLC SNAPSHOT(CH1616N)

 View more versions of this product

Catalog No. 14-037-771

  Check Availability
Add to Cart

Required Accessories : Luminometer

    Specifications

    Shelf Life 15 Months
    For Use With (Equipment) Charm NovaLum and Firefly 2 luminometer
    Required Accessories Luminometer
    Storage Requirements 2° to 8°C

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit