Learn More
Hygiena SnapShot™ Universal ATP Surface Test
Designed to be compatible with luminometers of most manufacturers
Manufacturer:  Hygiena SNAPSHOT (SPXL1333)
 View more versions of this product
Catalog No. 14-037-772
Add to cart
Requires : Luminometer
This item is not returnable. View return policy
Description
Description
Each version of Snapshot is designed to replicate the ATP reaction by generating the same light output as the device it is replacing; therefore eliminating the need to change current pass/fail threshold settings on the luminometer or within an established Quality Control program.
  • Unique, liquid-stable reagent provides unprecedented accuracy and reproducibility
  • Simple, quick and user-friendly
  • Reduced cost as compared to other devices intended for the luminometer
  • Tolerant to temperature abuse
  • Test wet or dry surfaces and liquid samples
Convenient design
  • Easy-to-use, all-in-one device
  • Snap-Valve™ technology; just snap and squeeze
  • Premoistened swab bud
  • Self-contained, liquid-stable reagent
  • Write-on swab label
Specifications
Specifications
2° to 8°C
BioTrace Uni-Lite NG luminometer
Luminometer
Safety and Handling
Safety and Handling
ShelfLife : 15 Months