Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygiena Transport-Swab

$194.00 - $208.50

Specifications

For Use With (Application) Ideal sampling device for customers looking for an environmental sample collection device
Includes Pre-filled Letheen broth, 10mL
Product Type Transport Swab
Shelf Life 18 Months
Sterility Sterile
View More Specs

Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
HTS10
View Documents Promotion Details
Hygiena
HTS10
Pre-filled Letheen broth, 10mL
Pack of 100 for $208.50
Add to cart
 
HTS4
View Documents Promotion Details
Hygiena
HTS4
Pre-filled Letheen broth, 4mL
Pack of 100 for $194.00
Add to cart
 
Description

Description

Specifications

Specifications

Ideal sampling device for customers looking for an environmental sample collection device
Transport Swab
Sterile
Rayon
Pre-filled Letheen broth, 10mL
18 Months
2 to 8°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit