Learn More
Hygiena Watershot™ Universal ATP Water Test
Designed to help companies conduct water testing with a luminometer other than a Hygiena ATP System
Manufacturer:  Hygiena WATERSHOT (WSPXL1333)
Catalog No. 14-037-775
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Requires : Luminometer
Description
Description
The 100 μL dipper tip, combined with a unique liquid-stable reagent, provides more accurate sample collection and reproducible results than tests that use a swab bud and freeze dried enzymes.
Watershot comes in two different configurations.
  • Simple, quick and user-friendly in the lab or out in the field
  • Total ATP test measures both microbial and nonmicrobial ATP
  • Unique liquid-stable reagent provides unprecedented accuracy and reproducibility—no reagent wastage
  • Consistent sample collection and reliable results
  • Tolerant to temperature abuse
  • Collects 100 μL of sample—no pipetting required
  • No need to change Pass/Fail setting; Watershot works exactly the same as the test it is replacing
Convenient design
  • Snap-Valve™ technology; just snap and squeeze
  • Honeycomb-shaped collection tip
  • Self-contained, liquid-stable reagent
  • Write-on swab label
  • Compatible with 3M Clean-Trace
Specifications
Specifications
2° to 8°C
3M Clean-Trace™
Luminometer
Safety and Handling
Safety and Handling
ShelfLife : 15 Months