Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYI Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548849

Catalog No. PA548849

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Catalyzes the reversible isomerization between hydroxypyruvate and 2-hydroxy-3-oxopropanoate (also termed tartronate semialdehyde).
Specifications

Specifications

HYI
Polyclonal
PBS with 0.09% sodium azide
Q5T013
Putative hydroxypyruvate isomerase, Endothelial cell apoptosis protein E-CE1, HYI
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
81888
Western Blot
Unconjugated
HYI
Liquid
HYI
KLH conjugated synthetic peptide between 47-73 amino acids from the N-terminal region of human HYI.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit