Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HYLS1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30514820UL
Description
HYLS1 Polyclonal specifically detects HYLS1 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
HYLS1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
219844 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunofluorescence | |
Unconjugated | |
FLJ32915HLS, hydrolethalus syndrome 1, hydrolethalus syndrome protein 1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 70-299 of human HYLS1 (NP_659451.1). | |
20 μL | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction