Learn More
Ansell™ Hynit™ 32-105 Series Blue Nitrile Medium Weight Gloves
Special nitrile formulation unites excellent resistance to abrasion with remarkable flexibility. Liquid-repellent impregnation repels oil, grease and dirt.
$18.41
Specifications
Glove Cuff Style Slip-on
View More Specs
Products 3
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
11-391-131B
View Documents
Ansell™
32105 9 208003
9 Dozen of 12 for $18.41
Case of 12 DZ for $220.90
Add to cart
11-391-131A
View Documents
Ansell™
32105 8 208002
8 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
11-391-131C
View Documents
Ansell™
32105 10 208006
10 This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Ideal for general plant use, assembly inspection, receiving, and light to medium fabrication and materials handling.
  • Nitrile-impregnated fabric ensures excellent abrasion and snag resistance
  • Feature clute cut and interlock knit liner for superior dry grip and excellent dexterity
  • Gloves are liquid repellent and machine washable so they will not shrink, stiffen, or lose their grip
  • Lining will not ball up
  • Color-coded cuffs for easy size identification and size mating after laundering
  • Available with solid and air-conditioned backs
Specifications
Specifications
Slip-on