Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hyoscyamine Sulfate, USP, 98.5-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
1 kg
Packaging:
Amber Glass Bottle
6835-16-1
C34H52N2O12S
712.85
BXSVDJUWKSRQMD-YBRNEGGZSA-N
bis(8-methyl-8-azabicyclo[3.2.1]octan-3-yl (2S)-3-hydroxy-2-phenylpropanoate) sulfuric acid dihydrate
O.O.OS(O)(=O)=O.CN1C2CCC1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1.CN1C2CCC1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1
6835-16-1
C34H52N2O12S
712.85
BXSVDJUWKSRQMD-YBRNEGGZSA-N
bis(8-methyl-8-azabicyclo[3.2.1]octan-3-yl (2S)-3-hydroxy-2-phenylpropanoate) sulfuric acid dihydrate
O.O.OS(O)(=O)=O.CN1C2CCC1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1.CN1C2CCC1CC(C2)OC(=O)[C@H](CO)C1=CC=CC=C1

HY136, 6835-16-1, (C17H23NO3)2•H2SO4•2H2O

Spectrum™ Hyoscyamine Sulfate, USP is used to control muscle spasms in the bladder, kidneys, or digestive tract, and to reduce stomach acid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Amber Glass Bottle
Quantity 100 g
Ignition Residue 0.001
Specific Rotation -24° to -29°
CAS Max % 1
UN Number UN1544
Percent Purity 98.5 to 100.5%