Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYOU1 Monoclonal Antibody (HL2006), Invitrogen™

Rabbit Monoclonal Antibody

Supplier:  Thermo Scientific MA547254

Encompass

Catalog No. PIMA547254


Add to Cart

Description

Description

Store as concentrated solution. Centrifuge briefly prior to opening vial.

HYOU1 Monoclonal antibody specifically detects HYOU1 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HYOU1
Monoclonal
1 mg/mL
PBS with no preservative
Q9Y4L1
HYOU1
Recombinant fragment of human ORP150
100 μL
Primary
Human
Antibody
IgG
Western Blot
HL2006
Unconjugated
HYOU1
140 kDa Ca(2+)-binding protein; 150 kDa oxygen-regulated protein; 150-kDa oxygen regulated protein; 170 kDa glucose regulated protein GRP170; 170 kDa glucose-regulated protein; AI415631; Cab140; calcium binding protein 140; calcium binding protein, 140 kDa; CBP-140; DKFZp686N08236; FLJ94899; FLJ97572; Grp 170; Grp170; GRP-170; HSP 12A; HSP12A; HYOU1; hypoxia up-regulated 1; hypoxia up-regulated protein 1; Orp 150; Orp150; ORP-150; oxygen regulated protein (150kD)
Rabbit
Protein A
RUO
10525
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit