Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HYOU1 Mouse anti-Human, Clone: 6G7-2H5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527643

Catalog No. PIMA527643


Add to cart

Description

Description

The protein encoded by this gene belongs to the heat shock protein 70 family. This gene uses alternative transcription start sites. A cis-acting segment found in the 5 ft. UTR is involved in stress-dependent induction, resulting in the accumulation of this protein in the endoplasmic reticulum (ER) under hypoxic conditions. The protein encoded by this gene is thought to play an important role in protein folding and secretion in the ER. Since suppression of the protein is associated with accelerated apoptosis, it is also suggested to have an important cytoprotective role in hypoxia-induced cellular perturbation. This protein has been shown to be up-regulated in tumors, especially in breast tumors, and thus it is associated with tumor invasiveness. This gene also has an alternative translation initiation site, resulting in a protein that lacks the N-terminal signal peptide. This signal peptide-lacking protein, which is only 3 amino acids shorter than the mature protein in the ER, is thought to have a housekeeping function in the cytosol. In rat, this protein localizes to both the ER by a carboxy-terminal peptide sequence and to mitochondria by an amino-terminal targeting signal.
Specifications

Specifications

HYOU1
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.1% sodium azide; pH 7.4
Q9Y4L1
ORP150, HSP12A, Hypoxia up regulated 1, Orp150, Glucose regulated 170, 150kDa oxygen regulated , 150 kDa oxygen regulated protein, 150 kDa oxygen-regulated protein, 170 kDa glucose regulated protein, 170 kDa glucose-regulated protein, DKFZp686N08236, FLJ94899, FLJ97572, glucose regulated protein 170, Grp 170, GRP-170, Grp170, HSP 12A, HSP12A, Hyou1, HYOU1_HUMAN, Hypoxia up regulated 1, hypoxia up-regulated 1 precursor, Hypoxia up-regulated protein 1, Hypoxia upregulated 1, Orp 150, ORP-150, Orp150, oxygen regulated protein (150kD)
Mouse
IgG2a
100 μg
-20°C
Primary
10525
Immunocytochemistry, Immunofluorescence, Western Blot
6G7-2H5
Unconjugated
HYOU1
Liquid
HYOU1
Raised against a synthetic peptide of human GRP170
Protein G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit