Thermo Scientific™ HyperClean™ Syringe Filters
HyperClean Syringe Filters offer greater performance and sample throughput.
Manufacturer: Thermo Scientific™ 60183-201
 View more versions of this product
Catalog No. 03-170-341
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »
    Specifications
    Housing Material Medical grade virginal polypropylene
    Item Description PTFE; 4mm DIA; 0.45uL
    Pore Size 0.45μm
    Diameter 4mm
    Material PTFE
    Thermo Scientific™ HyperClean Syringe Filters offer greater performance and sample throughput.
    • Chemically resistant polypropylene housing minimizes contamination
    • Uniform filtration across membrane surface
    • Female Luer-Lok™ design ensures safe and secure connections
    • Membrane type and porosity clearly identified on each filter
  • Description & Specifications
Related Products
Product Content Feedback