Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

Manufacturer:  Thermo Scientific™ SH30646.02

 View more versions of this product

Catalog No. SH3064602


Add to cart

Description

Description

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

Tank Liner Key Features:

  • Available in sizes that fit into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Drums (SV3011X, SV50076, and SV50517 series)
  • Gusset design allows for a close fit to the tank
  • Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific BPCs
  • Supplied gamma irradiated
  • Customization options are available by contacting one of our Thermo Scientific Representatives today

Standard Tank Liners for your liquid applications available in-stock in the following configurations:

SH30647.01 to .03:

  • 50 to 200L Tank Liner
  • Support Container Needed: Design to fit into HyPerforma Flat Bottom PE Drums (SV50076 series)
  • Quantity: Each = 1 Liner

SH30647.04 to .07:

  • 50 to 500L Tank Liner
  • Support Container Needed: Designed to fit into Industry Standard Cylindrical Tanks (SV3011X series)
  • Quantity: Each = 1 Liner

SH30646 Series:

  • 50 to 200L Tank Liner with One Bottom Drain
  • Bottom Lineset 1: 72in. (182.9cm) of tubing leading to a Quick Disconnect (MPX series) Insert (male) connector
  • Support Container Needed: Designed to fit into HyPerforma Conical Bottom PE Drums (SV50517 series)

Ordering Information:

Supplied double polybagged and irradiated with Certificate of Analysis.

Recommended for:

  • Hydrations of powdered media and buffers
  • Pooling of non-sterile solutions and fluids
  • Non-aseptic collection of waste liquids from separation and purification systems

To order, use the part numbers listed in the above table.

Specifications

Specifications

Liner - Bottom drain, irradiated; 100L; 1 Port; MPX Insert (Bottom)
HyPerforma Conical Bottom PE Drums
1/2in. Quick Disconnect Insert (MPX series)
Irradiated
Impeller Sleeve with Quick Connect: C-Flex™ Tubing
100L
CX3-9 Film
Packages are placed in strong cardboard cartons with product and lot identification.
Liners are packaged and sealed in two independent outer layers while still in the Class 10,000/ISO Class 7 area.
Ambient
No
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit