Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum

Use this LLDPE drum with 3-D bioprocess container systems and tank liners.

Manufacturer:  Thermo Scientific™ SV50076.02

 View more versions of this product

Catalog No. SV5007602


Add to cart

Description

Description

Use Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum with 3-D bioprocess container systems and tank liners.

  • Includes lid and clamps
  • Available in cylindrical, conical and flat-bottom
  • 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port
Specifications

Specifications

Lid
17.63 in.
Bioprocessing containers
53cm
HyPerforma™ Flat-Bottom LLDPE Drum
50L
Flat-bottom outer support container for bioprocess bags; Port detail: 4 and 1.75 in.
45cm
20.75 in.
Lid and Clamps
50L
LLDEP (Linear Low-density Polyethylene)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit