Learn More
Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum
Use this LLDPE drum with 3-D bioprocess container systems and tank liners.
Manufacturer:  Thermo Scientific™ SV50076.03
 View more versions of this product
Catalog No. SV5007603
Add to cart
Description
Description
Use Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum with 3-D bioprocess container systems and tank liners.
  • Includes lid and clamps
  • Available in cylindrical, conical and flat-bottom
  • 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port
Specifications
Specifications
Flat-bottom outer support container for bioprocess bags; Port detail: 4 and 1.75 in.
45cm
79cm
17.63 in.
31.25 in.
100L