Learn More
Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum
Use this LLDPE drum with 3-D bioprocess container systems and tank liners.
$177.72 - $273.87
Specifications
Description Flat-bottom outer support container for bioprocess bags; Port detail: 4 and 1.75 in.
View More Specs
Products 3
Catalog Number Mfr. No. Description Capacity (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Description Capacity (Metric) Price Quantity & Availability  
SV5007602 Green Product View Documents Thermo Scientific™
SV50076.02
Flat-bottom outer support container for bioprocess bags; Port detail: 4 and 1.75 in. 50L Each for $177.72
Add to cart
SV5007603 Green Product View Documents Thermo Scientific™
SV50076.03
Flat-bottom outer support container for bioprocess bags; Port detail: 4 and 1.75 in. 100L Each for $249.17
Add to cart
SV5007604 Green Product View Documents Thermo Scientific™
SV50076.04
Flat-bottom outer support container for bioprocess bags; Port detail: 4 and 1.75 in. 200L Each for $273.87
Add to cart
Description
Description
Use Thermo Scientific™ HyPerforma™ Flat-bottom LLDPE Drum with 3-D bioprocess container systems and tank liners.
  • Includes lid and clamps
  • Available in cylindrical, conical and flat-bottom
  • 4in. bottom port allows greater variety of customization to bottom port of BPC; can be used in place of drum with 1.75in. port
Specifications
Specifications
Flat-bottom outer support container for bioprocess bags; Port detail: 4 and 1.75 in.