Learn More
Thermo Scientific™ HyperSep™ Acid Wash SPE Fixed Plate
A polar sorbent primarily used to retain analytes in non-polar matrices.
Manufacturer: Thermo Scientific™ 60307324
Description
Applications:
- Extraction of polar compounds from ground, waste and drinking water
- Extraction of polar compounds from food and beverages
Specifications
100 mg | |
HyperSep | |
1/pk |