Learn More
Thermo Scientific™ HyperSep™ Acid Wash SPE Fixed Plate
A polar sorbent primarily used to retain analytes in non-polar matrices.
$284.50
Specifications
Quantity 1/pk
View More Specs
Products 4
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
60-307-321
View Documents
Thermo Scientific™
60307321
10 mg
Each for $284.50
Add to cart
60-307-322
View Documents
Thermo Scientific™
60307322
25 mg
Each for $284.50
Add to cart
60-307-323
View Documents
Thermo Scientific™
60307323
50 mg
Each for $284.50
Add to cart
60-307-324
View Documents
Thermo Scientific™
60307324
100 mg
Each for $284.50
Add to cart
Description
Description
Applications:
  • Extraction of polar compounds from ground, waste and drinking water
  • Extraction of polar compounds from food and beverages
Specifications
Specifications
1/pk