Learn More
Thermo Scientific™ HyperSep™ Acid Wash SPE Removable Plate
A polar sorbent primarily used to retain analytes in non-polar matrices.
Manufacturer:  Thermo Scientific™ 60300834
 View more versions of this product
Catalog No. 60-300-834
Add to cart
Description
Description
Applications:
  • Extraction of polar compounds from ground, waste and drinking water
  • Extraction of polar compounds from food and beverages
Specifications
Specifications
100 mg
1/pk
HyperSep