Learn More
Thermo Scientific™ HyperSep™ Acid Wash SPE Removable Plate
A polar sorbent primarily used to retain analytes in non-polar matrices.
$284.50
Specifications
Quantity 1/pk
View More Specs
Products 4
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
60-300-831
View Documents
Thermo Scientific™
60300831
10 mg
Each for $284.50
Add to cart
60-300-832
View Documents
Thermo Scientific™
60300832
25 mg
Each for $284.50
Add to cart
60-300-833
View Documents
Thermo Scientific™
60300833
50 mg
Each for $284.50
Add to cart
60-300-834
View Documents
Thermo Scientific™
60300834
100 mg
Each for $284.50
Add to cart
Description
Description
Applications:
  • Extraction of polar compounds from ground, waste and drinking water
  • Extraction of polar compounds from food and beverages
Specifications
Specifications
1/pk