Learn More
Thermo Scientific™ HyperSep Carboxylic Acid (WCX) Fixed Well Plates
Weak cation exchange sorbent for extraction of strong bases
$321.00
Specifications
Product Line HyperSep
Quantity 1/pk
View More Specs
Products 4
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
Catalog Number Mfr. No. Bed Weight Price Quantity & Availability  
60-307-331
View Documents
Thermo Scientific™
60307331
10 mg
Each for $321.00
Add to cart
60-307-332
View Documents
Thermo Scientific™
60307332
25 mg
Each for $321.00
Add to cart
60-307-333
View Documents
Thermo Scientific™
60307333
50 mg
Each for $321.00
Add to cart
60-307-334
View Documents
Thermo Scientific™
60307334
100 mg
Each for $321.00
Add to cart
Description
Description
Applications:
  • Drugs and metabolites in biological fluids
  • Peptides in serum, plasma or biological fluids
  • Pesticides in water, food and beverages
  • Nucleic acids, nucleotides and surfactants
Specifications
Specifications
HyperSep
1/pk