Learn More
Thermo Scientific™ HyperSep™ Dispersive SPE Clean-up Products (QuEChERS)
Save time and achieve more accurate pesticide determinations using these pre-prepared dispersive SPE products for QuEChERS clean-up.
Manufacturer:  Thermo Scientific™ 60105215SP
 View more versions of this product
Catalog No. 03-251-519
Add to cart
Description
Description
QuEChERS Dispersive SPE methods offer a convenient and effective approach for extraction and clean-up of pesticide residues in food and other complex matrices. The robust procedure offers a number of compelling advantages: high recoveries, accurate results, high sample throughput, low solvent and glassware usage, reduced labor and bench space and lower reagent costs.
Thermo Scientific™ HyperSep Dispersive SPE Clean-Up Products are available in a range of pre-prepared sorbent combinations, each containing the proper sorbents for optimum clean-up of analytes extracted using QuEChERS methods.
Comprehensive range of Dispersive SPE products that contain the proper sorbents foroptimum extraction, clean-up and separation of analytes from complex matrices such as food using QuEChERS methods.
  • Save valuable time in preparation – products are supplied pre-prepared
  • Reduced variability due to consistency in products and rigorous quality control
  • Cleaner extracts from cleaner, pre-prepared products
  • Lower peak counts as a result of using pre-prepared products
Specifications
Specifications
900 mg anhydrous magnesium sulfate, 150 mg PSA
Quechers
15 mL
50
Pouch 900 mg MgSO4, 150 mg PSA
QuEChERS Method
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.