Thermo Scientific™ HyperSep™ Filter Plates, 40¦L Bed Volume Available on GSA/VA Contract for Federal Government customers only.
Get simple, effective clean-up of proteins, peptides, DNA, RNA and other biomolecules in small-scale samples with these 96-well filter plates.
Manufacturer: Thermo Scientific™ 60110509
 View more versions of this product
Catalog No. 03-250-589
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With (Equipment) Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
    For Use With Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
    Sorbent HyperSep SAX
    Type 96-well Filter Plate
    Particle Size 40 - 60µm
    Item Description SAX; 40uL bed volume
    Description SAX; 40uL bed volume
    Achieve simple, effective purification and separation of proteins, peptides, DNA, RNA and other biomolecules in small-scale, microgram-level samples with Thermo Scientific™ HyperSep™ Filter Plates. Available in a range of chromatographic materials, the convenient 96-well plates can be used under vacuum.
    These HyperSep Filter Plates are available with 40µL bed volumes.
    HyperSep SPE Cartridges and Plates
    HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
    The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
  • Description & Specifications
Related Products
Product Content Feedback