Thermo Scientific™ HyperSep™ Florisil Plates Available on GSA/VA Contract for Federal Government customers only.
Isolate polar compounds from nonpolar matrices with these Florisil SPE well plates, which are especially well-suited for extraction of chlorinated pesticides.
Manufacturer: Thermo Scientific™ 60300727
 View more versions of this product
Catalog No. 03-251-231
  Check Availability
Add to Cart
    Specifications
    For Use With (Equipment) Isolation of Polar Compounds from Non-polar Matrices
    For Use With Isolation of Polar Compounds from Non-polar Matrices
    Sorbent HyperSep Magnesia-loaded Silica Gel
    Type 96-well Plate
    Particle Size 40 - 60µm
    Item Description Florisil 50mg/1mL 96 Removable Well Plate, 1pk
    Description Florisil 50mg/1mL 96 Removable Well Plate, 1pk
    The use of SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
    Thermo Scientific™ HyperSep™ Florisil Plates are ideal for the isolation of polar compounds from nonpolar matrices. Typical applications include:
    • Extraction of pesticides using official methods
    • Extraction of polychlorinated biphenyls (PCBs) in transformer oil
    • Alcohol, aldehydes, amines and drugs
    Extremely polar in nature, HyperSep Florisil SPE 96-Well Plates and Individual Wells are especially well-suited for extraction of chlorinated pesticides. The magnesia-loaded silica gel offers rapid flow rates for large sample volumes.
    HyperSep SPE Cartridges and Plates
    Thermo Scientific™ HyperSep™ solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
    The Thermo Scientific™ HyperSep-96™ system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are either available in packs of 100 individual wells or in pre-assembled plates.
  • Description & Specifications
Related Products
Product Content Feedback