Thermo Scientific™ HyperSep™ Glass Blocks for Vacuum Manifold Available on GSA/VA Contract for Federal Government customers only.
Order the glass block, which is the vacuum chamber of this manifold, if you need a replacement.
Manufacturer: Thermo Scientific™ 60104254
 View more versions of this product
Catalog No. 60-104-254
  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.
    Specifications
    For Use With Glass Block Vacuum Manifold
    Description Glass Block, 24 Port Vacuum Manifold, 1pk
    Order the Thermo Scientific™ HyperSep™ Vacuum Manifold Glass Block if you need a replacement. The vacuum chamber for this manifold is a glass block that allows you to see the extraction process and provides easy cleaning. The manifold enables sample preparation using HyperSep solid phase extraction (SPE) cartridges.
    HyperSep Glass Block Manifolds are durable, chemically resistant units designed to provide years of trouble-free extractions.
    Polypropylene racks are highly resistant to chemical degradation and abuse, and are supplied according to the number of positions on the manifold lid
    Vacuum control valve allows optimization of sample flow rates during extraction
    Test tube racks allow the use of 12-13mm or 16mm disposable test tubes
    Vacuum gauge and bleed valve enables easy monitoring of the vacuum process
    Corner leg supports prevent damage to the luers and tips when the lid is not being used
    HyperSep SPE Cartridges and Plates
    HyperSep solid phase extraction cartridges offer reproducible, reliable sample preparation in a traditional format. Available in a wide range of chemistries and bed weights, the cartridges are ideal for large samples and conform to industry standard configurations, allowing efficient routine processing by vacuum, by positive pressure or centrifugation.
    The HyperSep-96 system incorporates traditional solid phase extraction sorbents into 96-well plate format providing fast analysis without sacrificing accuracy or reproducibility. The innovative design of HyperSep-96 well plates provides flexibility for both method development and scale adjustment to 96-well without method revalidation. Well plates are customizable, allowing any arrangement of wells with differing sorbent chemistries for simultaneous processing. HyperSep-96 products are available in packs of 100 individual wells and in pre-assembled plates.
  • Description & Specifications
Related Products
Product Content Feedback