Learn More
Thermo Scientific™ HyperSep™ HPLC Columns for Online SPE Available on GSA/VA Contract for Federal Government customers only.
Clean-up and concentrate target analytes in minutes with these columns combining SPE chemistries with traditional HPLC column fittings and hardware.
Manufacturer:  Thermo Scientific™ 60312501
 View more versions of this product
Catalog No. 03-150-340
  Check Availability
Add to cart
Description
Description
Clean-up and concentrate target analytes in minutes with Thermo Scientific™ HyperSep™ Online SPE Products. Available in several convenient formats, the columns utilize popular HyperSep SPE chemistries. When used with conventional HPLC systems and appropriate switching valves, they provide:
  • Effective removal of contaminants, such as proteins, from samples
  • Fast, effective clean-up and concentration of target compounds
HyperSep HPLC Columns for Online SPE combine SPE chemistries with traditional HPLC column fittings and hardware.
HyperSep HPLC Columns for Online SPE are available with the following HyperSep SPE chemistries:
HyperSep™ Retain™ PEP — for retention of polar and nonpolar analytes
HyperSep Retain-CX — for retention of basic and nonpolar analytes
HyperSep Retain-AX — for retention of acidic and nonpolar analytes
HyperSep™ Hypercarb™ — for retention of extremely polar analytes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Online SPE
HyperSep Hypercarb
HyperSep Hypercarb; 2.1mm ID
Online SPE
Online HPLC Column
2.1mm ID
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.