Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ HyperSep™ Polystyrene Lab Plates
Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.
Supplier: Thermo Scientific™ 60110205
Description
Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include:
- Tissue culture and separation of products
- Concentration of small-scale samples
- Sample clean-up
- Collection of sample after chromatography
Specifications
C18 plus Hypercarb; Polystyrene | |
HyperSep | |
96-Well Plate |
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules | |
Hypercarb and HyperSep C18 (Mix Mode) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction