Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ HyperSep™ Polystyrene Lab Plates Available on GSA/VA Contract for Federal Government customers only.

Get simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules with these 96-well lab plates.

Manufacturer:  Thermo Scientific™ 60110206

 View more versions of this product

Catalog No. 03-250-559


  Check Availability
Add to cart

Specifications

Description

Available in a range of chromatographic materials, Thermo Scientific™ HyperSep™ Polystyrene Lab Plates enable simple, convenient purification and sample preparation for proteins, DNA, RNA and other biomolecules. In this 96 well plate format, media is embedded at the bottom of the plate. Typical applications include: 

  • Tissue culture and separation of products
  • Concentration of small-scale samples
  • Sample clean-up
  • Collection of sample after chromatography

The convenient 96-well HyperSep Lab Plates can be processed manually or using a liquid-handling robot. They are not suitable for use with a vacuum.
Description & Specifications

Specifications

Zirconium Dioxide; Polystyrene
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
96-well Plate
Purification and Sample Preparation of Proteins, DNA, RNA and Other Biomolecules
HyperSep Zirconium Dioxide
Zirconium Dioxide; Polystyrene
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit