Learn More
Thermo Scientific™ HyperSep? Pressure Manifold Adapter Plates Available on GSA/VA Contract for Federal Government customers only.
Use the adapter plate that matches the size of solid phase extraction (SPE) cartridges used with the Thermo Scientific™ HyperSep™ Positive Pressure Manifold.
Manufacturer:  Thermo Scientific™ 60104271
 View more versions of this product
Catalog No. 60-104-271
  Check Availability
Add to cart
Specifications
Description
Use the Thermo Scientific? HyperSep? Positive Pressure Manifold Adapter Plate that matches the size of solid phase extraction (SPE) cartridges used. The HyperSep Positive Pressure Manifold can hold up to 48 SPE cartridges of various sizes: 1, 3, 6, and 10/15mL.
The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes. This SPE manifold provides a quick, simple and robust way to prepare multiple samples for analysis by chromatography.
See related products if you need replacements for any of these HperSep Positive Pressure Manifold accessories.
Description & Specifications
Specifications
Adapter Plate for 6mL Cartridges for PP Manifold, 1pk
HyperSep Positive Pressure Manifold