Thermo Scientific™ HyperSep™ Servo AMP Columns and Well Plates

An optimized SPE solution for extraction of AMP and associated metabolites.

Manufacturer: Thermo Scientific™ 60111606C

 View more versions of this product

Catalog No. 60-111-606C

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    For Use With (Equipment) Analysis of Drugs of Abuse
    Type Extraction Column
    Bed Weight 1g
    For Use With Analysis of Drugs of Abuse
    Sorbent HyperSep Servo AMP
    Description 1g/6mL SPE Column 30Pk
    Volume (Metric) 6mL

    Thermo Scientific™ HyperSep Servo AMP Columns and Well Plates provide an optimized SPE solution for extraction of AMP and associated metabolites.

    Greater performance helps provide improved confidence in results. HyperSep Servo+ and HyperSep Servo SPE product ranges remove lack of confidence in your sample preparation methodology by supplying an optimized extraction solution for analysis of Drugs of Abuse.

    HyperSep Servo+ and HyperSep Servo offer high levels of reproducibility and cleanliness of sample extract essential for dealing with complex biological sample matrices which are often associated with analysis of Drugs of Abuse. HyperSep Servo+ provides the additional benefit of greater selectivity, higher loading capacities and robustness.

    • Ready-to-use, optimized solutions
    • Greater reproducibility and cleanliness of extract
    • Greater confidence in your results

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit