Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Extraction Tips, 10-200¦L Available on GSA/VA Contract for Federal Government customers only.
Get faster sample preparation with minimal sample loss with these SPE micropipette tips and achieve separation in volumes as low as 100nL.
Manufacturer: Thermo Scientific™ 60109420
 View more versions of this product
Catalog No. 03-250-447
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    For Use With (Equipment) Microscale Solid Phase Extraction
    For Use With Microscale Solid Phase Extraction
    Item Description Strong Anion Exchanger; 10-200uL Vol.
    Sorbent HyperSep Poros SAX
    Type Spintip
    Particle Size 40 - 60µm
    Description Strong Anion Exchanger; 10-200uL Vol.
    Get faster sample preparation with minimal sample loss with the revolutionary Thermo Scientific™ HyperSep™ SpinTip Microscale SPE Tips and achieve separation in volumes as low as 100nL.
    Features:
    Ideal for small sample volumes; features a 1 to 2µm wide slit at the bottom that permits liquid to pass through while retaining the chromatographic material (20 to 30µm).
    • [bullet] Filter-free design to eliminate dead volume
    • [bullet] No contamination from the supporting matrix
    • [bullet] Available in many popular chemistries
    Applications include:
    • [bullet] Mass spectrometry
    • [bullet] Desalting
    • [bullet] Protein purification
    • [bullet] MALDI
    • [bullet] Electrophoresis
    • [bullet] HPCE
    • [bullet] HPLC
    • [bullet] CEC
  • Description & Specifications
Related Products
Product Content Feedback