Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 1-10µL Available on GSA/VA Contract for Federal Government customers only.
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Manufacturer: Thermo Scientific™ 60109203
 View more versions of this product
Catalog No. 03-250-414
  Check Availability
Add to Cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
    Specifications
    Item Description BioBasic C4, 1 to 10µL Volume
    Particle Size
    Description BioBasic C4, 1 to 10µL Volume
    For Use With (Equipment) Microscale Solid Phase Extraction
    For Use With Microscale Solid Phase Extraction
    Sorbent BioBasic C4
    Type Tip
    Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
    Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
  • Description & Specifications
Related Products
Product Content Feedback