Learn More
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 10-200µL Available on GSA/VA Contract for Federal Government customers only.
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips.
Manufacturer:  Thermo Scientific™ 60109218
 View more versions of this product
Catalog No. 03-250-551
Add to cart
Description
Description
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
HyperSep Microscale Solid Phase Extraction Tips are ideal when you're working with small sample volumes. Typical applications include:
Mass spectrometry
Desalting
Protein purification
MALDI
Electrophoresis
HPCE
HPLC
CEC
Tips are available in 1-10µL capacity and 10-200µL capacity.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Zirconium Dioxide; 10 to 200uL Volume
Microscale Solid Phase Extraction
Tip
Microscale Solid Phase Extraction
HyperSep Zirconium Dioxide
Zirconium Dioxide; 10 to 200uL Volume
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.