Learn More
Thermo Scientific™ Hypersil™ BDS C18 Columns Available on GSA/VA Contract for Federal Government customers only.
For a robust, general-purpose column to use in a wide range of applications, choose Thermo Scientific? Hypersil? BDS C18 HPLC Columns.
Manufacturer:  Thermo Scientific™ 28105124630
 View more versions of this product
Catalog No. 28105124630
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
For a robust, general-purpose column to use in a wide range of applications, choose Thermo Scientific? Hypersil? BDS C18 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
For more challenging separations that require improved chromatographic performance, use Thermo Scientific? Hypersil GOLD? columns as an alternative.
  • Original Hypersil columns from the manufacturer
  • Base deactivated with minimal residual silanol activity
  • Exceptional reproducibility
  • Economical, general purpose columns
Ordering Information:
Other column dimensions are available. Please call Customer Service for more information.
Description & Specifications
Specifications
11%
Reversed Phase
Yes
400bar
5μm
130Å
60°C
C18
Analytical Column
4.6mm
125mm
Silica, Spherical Fully Porous Base Deactivated
2 to 8
170m²/g
L1