Thermo Scientific™ Hypersil™ BDS C8 LC Columns Available on GSA/VA Contract for Federal Government customers only.

For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo Scientific™ Hypersil™ BDS C8 HPLC Columns.

Manufacturer: Thermo Scientific™ 28203154630

 View more versions of this product

Catalog No. 03-050-526

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Pore Size 130Å
    Column Format Analytical Column
    Diameter (Metric) 4.6mm
    Column Type Reversed Phase
    Max. Pressure 400bar
    Packing Material Spherical, fully porous base deactivated silica
    Particle Size 3µm
    Temperature 60°C
    pH 2 to 8
    For Use With HPLC
    Stationary Phase C8
    Surface Area 170m²/g
    Length (Metric) 150mm
    Carbon Load 7%
    USP Type L7
    Endcapped Yes

    For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo Scientific™ Hypersil™ BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.

    For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

    • Excellent choice for acids, bases and neutral compounds
    • Less retentive than C18 for faster analyses
    • Robust columns with long column lifetimes
    • Very reliable for routine assays

    Ordering Information:

    Other column dimensions are available. Please call Customer Service for more information.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit