Learn More
Thermo Scientific™ Hypersil™ Phenyl-BDS LC Columns
For a base-deactivated column with unique selectivity for aromatic and slightly polar compounds, use Thermo Scientific™ Hypersil™ Phenyl-BDS LC Columns.
Manufacturer:  Thermo Scientific™ 28903014001
 View more versions of this product
Catalog No. 28-903-014001
Add to cart
Description
Description
As a base-deactivated column with unique selectivity for aromatic and slightly polar compounds, you can use Thermo Scientific™ Hypersil™ Phenyl-BDS LC Columns as an alternative selectivity to C18 or C8 phases. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.
For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ columns as an alternative.
Convenience, economy and effective protection.
  • 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific™ UNIGUARD™ direct connection and stand alone holders
Specifications
Specifications
5%
Reversed Phase
Yes
400bar
3 µm
130 Å
60°C
Phenyl
Guard Cartridge, 4 Pk.
4 mm
10 mm
Silica, Spherical Fully Porous Base Deactivated
2 to 8
170 m²/g
L11
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.