Thermo Scientific™ Hypersil GOLD™ Amino LC Columns Available on GSA/VA Contract for Federal Government customers only.

Achieve outstanding peak shape and sensitivity in weak anion exchange, reversed phase and normal phase modes with Hypersil GOLD Amino LC Columns.

Manufacturer: Thermo Scientific™ 25703052130

 View more versions of this product

Catalog No. 25-703-052130

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Column Format Analytical Column
    Diameter (Metric) 2.1mm
    Column Type Normal Phase
    Packing Material Spherical, fully porous ultrapure silica
    pH 2 to 8
    For Use With HILIC
    Stationary Phase Amino
    Surface Area 220m²/g
    Type Analytical Column
    Length (Metric) 50mm
    USP Type L8
    Endcapped Yes
    Max. Pressure 400bar
    Particle Size 3µm
    Temperature 60°C
    Pore Size 175Å
    Carbon Load 0.02

    Achieve excellent chromatographic separations in weak anion exchange, reversed phase and normal phase modes with Thermo Scientific™ Hypersil GOLD™ Amino LC Columns. These columns, featuring a high-performance aminopropyl phase, provide outstanding peak shape and sensitivity and are available with a 1.9µm particle size for high speed, high efficiency separations.

    Excellent Chromatographic Properties in Three Modes
    • Retains anions and organic acids in weak anion exchange mode when used with common buffers and an organic modifier
    • Excels for carbohydrate analysis in reversed phase mode
    • Offers an alternative selectivity to silica columns in normal phase chromatography
    About the Hypersil GOLD Family of HPLC Columns
    Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
    • Excellent peak symmetry
    • Narrow peaks for outstanding efficiency
    • Enhanced sensitivity and improved resolution
    • Exceptional reproducibility for reliable chromatography, column after column
    • Solutions for high-throughput screening1.9, 3 and 5µm particle sizes for all your separation needs

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit