Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersil GOLD™ AX LC Columns

Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns.

Manufacturer:  Thermo Scientific™ 26105104630

 View more versions of this product

Catalog No. 26-105-104630


Add to cart

Description

Description

Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns. The weak anion-exchange phase, created by a polymeric amine ligand bonded to highly pure base-deactivated silica, separates multiple charged analytes. The bonded phase offers additional chromatographic options, including hydrophilic interaction chromatography (HILIC), which is ideal for the separation of sugars and other highly polar species.

Excellent Chromatographic Performance
  • Outstanding peak shape and selectivity
  • Superb stability under demanding pH conditions
  • Higher efficiency than polymer-based ion exchange columns
  • Improved speed with 1.9µm particle size columns
About the Hypersil GOLD Family of HPLC Columns



Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5µm particle sizes for all your separation needs
Specifications

Specifications

6%
Ion Exchange
No
400bar
5 µm
175 Å
60°C
Polyethyleneimine
Analytical Column
4.6 mm
100 mm
Silica, Spherical Fully Porous Ultrapure
2 to 8
220 m²/g
Analytical Column
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.