Thermo Scientific™ Hypersil GOLD™ AX LC Columns Available on GSA/VA Contract for Federal Government customers only.

Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns.

Manufacturer: Thermo Scientific™ 26102102130

 View more versions of this product

Catalog No. 26-102-102130

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Column Format Analytical Column
    Diameter (Metric) 2.1mm
    Temperature 60°C
    For Use With HILIC
    Format Analytical Column
    USP Type N/A
    Column Type Ion Exchange
    Endcapped No
    Max. Pressure 1250bar
    Packing Material Spherical, fully porous ultrapure silica
    Particle Size 1.9µm
    pH 2 to 8
    Stationary Phase Polyethyleneimine
    Surface Area 220m²/g
    Length (Metric) 100mm
    Carbon Load 0.06
    Pore Size 175Å
    Type Analytical Column
    Diameter 2.1mm

    Efficiently separate smaller proteins, peptides, anionic species and polar molecules with Thermo Scientific™ Hypersil GOLD™ AX LC Columns. The weak anion-exchange phase, created by a polymeric amine ligand bonded to highly pure base-deactivated silica, separates multiple charged analytes. The bonded phase offers additional chromatographic options, including hydrophilic interaction chromatography (HILIC), which is ideal for the separation of sugars and other highly polar species.

    Excellent Chromatographic Performance
    • Outstanding peak shape and selectivity
    • Superb stability under demanding pH conditions
    • Higher efficiency than polymer-based ion exchange columns
    • Improved speed with 1.9µm particle size columns 
    About the Hypersil GOLD Family of HPLC Columns

    Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.
    • Excellent peak symmetry
    • Narrow peaks for outstanding efficiency
    • Enhanced sensitivity and improved resolution
    • Exceptional reproducibility for reliable chromatography, column after column
    • Solutions for high-throughput screening
    • 1.9, 3 and 5µm particle sizes for all your separation needs 

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit