Thermo Scientific™ Hypersil GOLD™ C4 Analytical HPLC Columns, 3µm

Achieve faster separations for very hydrophobic analytes with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns.

Manufacturer: Thermo Scientific™ 25503051030

 View more versions of this product

Catalog No. 25-503-051030

  Check Availability
Add to Cart

    Specifications

    Pore Size 175Å
    Stationary Phase Hypersil Gold C4
    Diameter (Metric) 1mm
    Column Type Reversed-Phase
    Particle Size 3µm
    For Use With HPLC
    Length (Metric) 50mm

    Feature lower hydrophobicity than C18 or C8 and are recommended for very hydrophobic analytes with Thermo Scientific™ Hypersil GOLD™ 3µm C4 Analytical HPLC Columns. Based on high purity silica, Hypersil GOLD is a state-of-the-art family of columns designed to meet the challenges of modern chromatography.

    Similar Selectivity, Less Retention Than C18 or C8 Columns

    • Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
    • Ideal for analysis of hydrophobic polypeptides and proteins

    Superior Performance

    • Highly symmetrical peak shapes 
    • 1.9µm particle size columns improve speed and efficiency

    About the Hypersil GOLD Family of HPLC Columns

    Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.

    • Excellent peak symmetry
    • Narrow peaks for outstanding efficiency
    • Enhanced sensitivity and improved resolution
    • Exceptional reproducibility for reliable chromatography, column after column
    • Solutions for high-throughput screening
    • 1.9, 3 and 5µm particle sizes for all your separation needs

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit