Thermo Scientific™ Hypersil GOLD™ C4 LC Columns Available on GSA/VA Contract for Federal Government customers only.

Featuring a lower hydrophobicity than C18 or C8 columns, Thermo Scientific™ Hypersil GOLD™ C4 columns provide outstanding sensitivity and high efficiency.

Manufacturer: Thermo Scientific™ 25505152130

 View more versions of this product

Catalog No. 25-505-152130

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Column Format Analytical Column
    Diameter (Metric) 2.1mm
    Column Type Reversed Phase
    Particle Size 5µm
    For Use With ----
    Stationary Phase C4
    Packing Material Spherical, fully porous ultrapure silica
    Temperature 60°C
    Pore Size 175Å
    pH 2 to 8
    Surface Area 220m²/g
    USP Type L26
    Max. Pressure 400bar
    Length (Metric) 150mm
    Carbon Load 5%
    Endcapped Yes

    For very hydrophobic analytes, achieve faster separations with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns. Featuring a lower hydrophobicity than C18 or C8 columns, Hypersil GOLD C4 columns provide outstanding sensitivity and high efficiency.

    Similar selectivity, less retention than C18 or C8 columns

    • Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
    • Ideal for analysis of hydrophobic polypeptides and proteins
    Superior Performance
    • Highly symmetrical peak shapes
    • 1.9µm particle size columns improve speed and efficiency
    About the Hypersil GOLD family of HPLC columns

    Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC, or normal phase chromatography.

    • Excellent peak symmetry
    • Narrow peaks for outstanding efficiency
    • Enhanced sensitivity and improved resolution
    • Exceptional reproducibility for reliable chromatography, column after column
    • Solutions for high-throughput screening
    • 1.9, 3, and 5µm particle sizes for all your separation needs

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit