Learn More
Thermo Scientific™ Hypersil GOLD™ Cyano LC Columns, 3µm particle size, 250mm, 2.1mm I.D.
Alternate selectivity with lower hydrophobicity. Use these HPLC columns for normal and reversed phase separations.
Manufacturer:  Thermo Scientific™ 25805013001
 View more versions of this product
Catalog No. 25805013001
Add to cart
Description
Description
Alternate selectivity with lower hydrophobicity. Use Thermo Scientific™ Hypersil Gold™ Cyano LC Columns for normal and reversed phase separations.
  • Excellent peak shape
  • Outstanding sensitivity
  • Less retention for faster analysis
Specifications
Specifications
4%
Normal Phase
Yes
10 mm
5 µm
175 Å
60°C
L10
Guard Cartridge, 4 Pk.
3 mm
Uniguard Direct-connection and Stand Alone Holder
400bar
2 to 8
220 m²/g
Guard Cartridge, 4 Pk.
Cyano
Documents
Accessories
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.