Thermo Scientific™ Hypersil GOLD™ HILIC HPLC Columns, 3µm

Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Thermo Scientific™ Hypersil GOLD™ HILIC HPLC Columns.

Manufacturer: Thermo Scientific™ 26503101030

 View more versions of this product

Catalog No. 26-503-101030

  Check Availability
Add to Cart

    Specifications

    Column Type HILIC
    Particle Size 3µm
    Pore Size 100Å
    pH 2 - 8
    For Use With HILIC, ESI-MS
    Stationary Phase Hypersil Gold HILIC
    Surface Area 320m²/g
    Length (Metric) 100mm
    Carbon Load 6%
    Diameter (Metric) 1mm
    Endcapped Yes

    Retain highly polar and hydrophilic analytes, which are difficult to analyze with conventional C18 columns, using Thermo Scientific™ Hypersil GOLD™ HILIC HPLC Columns. These columns deliver straightforward separation without the need for sample derivatization or ion-pair reagents. Featuring outstanding peak shape and selectivity, the columns provide improved sensitivity for MS detection. With 1.9, 3 and 5µm particle sizes, Hypersil GOLD HILIC (hydrophilic interaction liquid chromatography) columns are ideal for applications within drug discovery, food testing and environmental analysis.

    About the Hypersil GOLD Family of HPLC Columns

    Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.

    • Excellent peak symmetry
    • Narrow peaks for outstanding efficiency
    • Enhanced sensitivity and improved resolution
    • Exceptional reproducibility for reliable chromatography, column after column
    • Solutions for high-throughput screening
    • 1.9, 3 and 5µm particle sizes for all your separation needs

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit