Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersil™ GOLD PFP Preparative HPLC Guard Columns

Protect Thermo Scientific™ Hypersil™ GOLD PFP Preparative HPLC Columns from sample debris and interfering substances.

Specifications

Carbon Load 8%
Endcapped No
Pore Size 175Å
Surface Area 220 m2/g
USP Type L43
View More Specs

Products 2
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
03-051-417
View Documents
Thermo Scientific™
25405039022
5 µm 10 mm 30 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
03-051-418
View Documents
Thermo Scientific™
25405039222
5 µm 21 mm 30 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Protect your preparative column against damage from sample debris and interfering substances. Extend column lifetime with Thermo Scientific™ Hypersil™ GOLD PFP Preparative Guard Cartridges. For use with Hypersil GOLD PFP Preparative Columns, which separate difficult-to-resolve mixtures of halogenated compounds and nonhalogenated polar compounds.

Features of Hypersil GOLD PFP Solid Phase

  • Hypersil GOLD PFP (perfluorinated phenyl) columns offer alternative selectivity: the fluorine atoms around the phenyl ring enhance pi-pi interactions with aromatic molecules
  • Alternative selectivity to C18
  • Extra retention for halogenated species
  • Selectvity for non-halogenated polar compounds
  • Excellent peak shape and sensitivity
Specifications

Specifications

8%
175Å
L43
5 µm
No
220 m2/g
30 mm
2 to 8
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit