Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Hypersil™ GOLD Preparative HPLC Guard Columns

Protect Thermo Scientific™ Hypersil™ GOLD Preparative HPLC Columns from sample debris and interfering substances.

Specifications

Carbon Load 10%
Endcapped Yes
Pore Size 175Å
Surface Area 220 m2/g
USP Type L1
View More Specs

Products 2
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
25005039022
View Documents
Thermo Scientific™
25005039022
5 µm 10 mm 30 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
25012039222
View Documents
Thermo Scientific™
25012039222
12 µm 20 mm 30 mm This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Protect your preparative column against damage from sample debris and interfering substances. Extend column lifetime with Thermo Scientific™ Hypersil™ GOLD Preparative HPLC Guard Cartridges. For use with Hypersil GOLD Preparative HPLC Columns, which provide exceptional peak shape and resolution for all analyte types.

  • Hypersil GOLD HPLC columns provide excellent peak shape for all analyte types
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitvity
  • Confidence in the accuracy and quality of analytical data
Specifications

Specifications

10%
175Å
L1
1 to 11
Yes
220 m2/g
30 mm
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit