Thermo Scientific™ Hypersil™ MOS-2 C8 LC Columns Available on GSA/VA Contract for Federal Government customers only.

Use these C8 columns for reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids and environmental samples.

Manufacturer: Thermo Scientific™ 30305254030

 View more versions of this product

Catalog No. 30305254030

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.


    Specifications

    Column Format Analytical Column
    Diameter (Metric) 4mm
    Column Type Reversed-Phase
    Temperature 60°C
    Surface Area 170m²/g
    Certifications/Compliance Manufactured under ISO 9001-2000 quality guidelines.
    USP Type L7
    Max. Pressure 400bar
    Packing Material Spherical, fully porous Type A silica
    Particle Size 5µm
    Pore Size 120Å
    pH 2 - 8
    For Use With ----
    Stationary Phase C8
    Length (Metric) 250mm
    Carbon Load 6.5%
    Endcapped No

    For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS-2 C8 HPLC Columns.

    Hypersil stationary phases are recognized as an HPLC industry standard. A proven effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.

    For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.

    • Reliable, less retentive phase
    • Long column lifetimes, even under basic conditions
    • Manufactured under ISO 9001-2000 quality guidelines
    • Affordable

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit