Learn More
Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns
Use as a general-purpose C8 phase for acidic and neutral analytes.
$569.00 - $1359.00
Specifications
Max. Pressure | 5800 psi (400 bar) |
---|---|
pH | 2 to 8 |
Pore Size | 120 Å |
Surface Area | 170 m²/g |
Temperature | 60°C |
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
30-205-054630
|
Thermo Scientific™
30205054630 |
5 μm | 4.6 mm | 50 mm |
Each for $820.00
|
|
|||
03-050-127
|
Thermo Scientific™
30205154630 |
5 μm | 4.6 mm | 150 mm |
Each for $1,229.00
|
|
|||
30-205-254030
|
Thermo Scientific™
30205254030 |
5 μm | 4 mm | 250 mm |
Each for $1,317.00
|
|
|||
03-050-128
|
Thermo Scientific™
30205254630 |
5 μm | 4.6 mm | 250 mm |
Each for $1,359.00
|
|
|||
03-150-545
|
Thermo Scientific™
30205014001 |
5 μm | 4 mm | 10 mm |
Pack of 4 for $569.00
|
|
|||
03-260-199
|
Thermo Scientific™
30210204630 |
10 μm | 4.6 mm | 200 mm |
Each for $864.50
|
|
|||
Description
For reliable, affordable analysis of more hydrophobic analytes such as small peptides and proteins, pharmaceuticals, steroids, and environmental samples, use Thermo Scientific™ Hypersil™ MOS C8 HPLC Columns.
Recognized as an HPLC industry standard, Hypersil stationary phases are proven effective analytical tools. Classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 guidelines.
For separations requiring improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ C8 columns as an alternative.
- Reliable, less retentive phase
- Long column lifetimes, even under basic conditions
- Manufactured under ISO 9001-2000 quality guidelines
- Affordable
Ordering Information:
Other column dimensions are available. Also available in 10 μm. Please call Customer Service for more information.
Specifications
5800 psi (400 bar) | |
120 Å | |
60°C | |
C8 | |
6.5% |
2 to 8 | |
170 m²/g | |
L7 | |
Hypersil Classical | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.