Learn More
Thermo Scientific™ Hypersil™ ODS C18 Columns Available on GSA/VA Contract for Federal Government customers only.
Choose Thermo Scientific™ Hypersil™ ODS C18 HPLC Columns for a general purpose C18 phase to use in a broad range of applications.
Manufacturer:  Thermo Scientific™ 30103254030
 View more versions of this product
Catalog No. 30103254030
  Check Availability
Add to cart
Disclaimers:
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Description
Choose Thermo Scientific? Hypersil? ODS C18 HPLC Columns for a general purpose C18 phase to use in a broad range of applications.
Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, the classical Hypersil phases are well established and referenced in many HPLC methods worldwide. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, we recommend the use of Thermo Scientific? Hypersil GOLD? columns as an alternative.
An excellent C18 phase for a broad range of applications.
  • High efficency
  • Proven reproducibility
  • Exceptionally reliable
Description & Specifications
Specifications
10%
Reversed Phase
Yes
400bar
3μm
120Å
60°C
C18
Analytical Column
4mm
250mm
Silica, Spherical Fully Porous Type A
2 to 8
170m²/g
L1