Learn More
Thermo Scientific™ Hypersil™ ODS (C18) HPLC Columns, 2/3um Particle Size Available on GSA/VA Contract for Federal Government customers only.
Separate a wide range of compounds, including nonpolar, moderately polar and lipophilic compounds like triglycerides.
Manufacturer:  Thermo Scientific™ 30203054630
 View more versions of this product
Catalog No. 03-150-539
Add to cart
Description
Description
Separate a wide range of compounds, including nonpolar, moderately polar and lipophilic compounds like triglycerides with Thermo Scientific™ Hypersil™ ODS (C18) HPLC Columns.
An excellent C18 phase for a broad range of applications.
  • High efficency
  • Proven reproducibility
  • Exceptionally reliable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.