Learn More
Thermo Scientific™ Hypersil™ Phenyl LC Columns Available on GSA/VA Contract for Federal Government customers only.
Effectively separate aromatic compounds and moderately polar compounds with the reversed-phase Thermo Scientific™ Hypersil™ Phenyl LC Columns.
Manufacturer:  Thermo Scientific™ 30905014001
 View more versions of this product
Catalog No. 03-050-556
Add to cart
Description
Description
Effectively separate aromatic compounds and moderately polar compounds with Thermo Scientific™ Hypersil™ Phenyl LC Columns.
Classical Hypersil stationary phases are recognized as an industry standard in HPLC. Proven as an effective analytical tool, classical Hypersil phases are well established and referenced in HPLC methods. Quality is assured by manufacture under ISO 9001-2000 quality guidelines.
For more challenging separations that require improved chromatographic performance, use Thermo Scientific™ Hypersil GOLD™ Phenyl columns as an alternative.
  • The 10mm design offers maximum protection with minimal increase in retention Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Available in packs of 4
Features of Phenyl Solid Phase
  • For the separation of certain aromatic compounds and moderately polar compounds
  • Retention characteristics similar to Hypersil MOS
  • Alternative selectivity to C8
Requires:
Thermo Scientific Uniguard™ direct connection or stand-alone holders
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
5%
Reversed Phase
No
10mm
5μm
120Å
60°C
Guard Cartridge, 4 PK
4mm
Uniguard Direct-connection and Stand Alone Holder
400bar
2 to 8
170m²/g
L11
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.